accessibility menu, dialog, popup

UserName

Viewing profile as user:
POHOOHPOHOOH

CAS RN 880-68-2

IUPAC Name: (4-phosphonophenyl)phosphonic acid
Synonyms: 4-phosphonophenylphosphonic acid
Molecular Weight (g/mol): 238.07
Molecular Formula: C6H8O6P2
InChi Key: JHDJUJAFXNIIIW-UHFFFAOYSA-N
SMILES: OP(O)(=O)C1=CC=C(C=C1)P(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results