accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 880166-18-7

IUPAC Name: 4-(thiophen-2-yl)oxane-4-carboxylic acid
Synonyms: 4-(thiophen-2-yl)oxane-4-carboxylic acid
Molecular Weight (g/mol): 212.26
Molecular Formula: C10H12O3S
InChi Key: VWYOZQWRYINSBP-UHFFFAOYSA-N
SMILES: OC(=O)C1(CCOCC1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results