accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 880361-73-9

IUPAC Name: 1-[(2-methyl-1,3-thiazol-4-yl)methyl]piperazine
Synonyms: 1-[(2-methyl-1,3-thiazol-4-yl)methyl]piperazine
Molecular Weight (g/mol): 197.3
Molecular Formula: C9H15N3S
InChi Key: RZWRPESYZFJDIU-UHFFFAOYSA-N
SMILES: CC1=NC(CN2CCNCC2)=CS1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results