accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNNNNN

CAS RN 88088-95-3

IUPAC Name: 1-[bis(1H-1,2,3-benzotriazol-1-yl)methyl]-1H-1,2,3-benzotriazole
Synonyms: 1-[bis(1,2,3-benzotriazol-1-yl)methyl]-1,2,3-benzotriazole
Molecular Weight (g/mol): 367.38
Molecular Formula: C19H13N9
InChi Key: VENLCXAGJRTBFL-UHFFFAOYSA-N
SMILES: C1=CC=C2N(N=NC2=C1)C(N1N=NC2=CC=CC=C12)N1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)