accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3

CAS RN 881-57-2

IUPAC Name: 5-formyl-2-methoxyphenyl acetate
Synonyms: 5-formyl-2-methoxyphenyl acetate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: ZVPGTXJXZIXWGR-UHFFFAOYSA-N
SMILES: COC1=C(OC(C)=O)C=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results