accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 881190-93-8

IUPAC Name: 3-(3-chloro-4-fluorophenyl)propanoic acid
Synonyms: 3-(3-chloro-4-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: XJSXYOSRQKEOSZ-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(Cl)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results