accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88145-89-5

IUPAC Name: 6-bromo-1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms: 6-bromo-1,3-dihydroquinazoline-2,4-dione
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: JZDVFUAHGLJVQG-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)NC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results