accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH2NBrOFFF

CAS RN 88149-49-9

IUPAC Name: 2,6-dibromo-4-(trifluoromethoxy)aniline
Synonyms: 2,6-dibromo-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 334.92
Molecular Formula: C7H4Br2F3NO
InChi Key: JBSWOEGXMADXOU-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(OC(F)(F)F)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results