accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3H3CCH3NOCH3

CAS RN 881656-35-5

IUPAC Name: 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 285.15
Molecular Formula: C16H20BNO3
InChi Key: MGEKTRDPBDREKY-UHFFFAOYSA-N
SMILES: COC1=CN=C2C=CC=C(B3OC(C)(C)C(C)(C)O3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results