accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COClOHO

CAS RN 882-09-7

IUPAC Name: 2-(4-chlorophenoxy)-2-methylpropanoic acid
Synonyms: fibrates clofibric acid regulipid regadrin 4-cpib pcpib chlorfibrinic acid clofibrin p-chlorophenoxyisobutyric acid PCIB
Molecular Weight (g/mol): 214.65
Molecular Formula: C10H11ClO3
InChi Key: TXCGAZHTZHNUAI-UHFFFAOYSA-N
SMILES: CC(C)(OC1=CC=C(Cl)C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)