CAS RN 882268-69-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ HIF-2α Translation Inhibitor
The HIF-2α Translation Inhibitor, also referenced under CAS 882268-69-1, controls the biological activity of HIF-2α. This small molecule/inhibitor is primarily used for Cell Structure applications.