accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 883-62-5

IUPAC Name: 3-methoxynaphthalene-2-carboxylic acid
Synonyms: 3-methoxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: RTBQQRFTCVDODF-UHFFFAOYSA-N
SMILES: COC1=C(C=C2C=CC=CC2=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results