accessibility menu, dialog, popup

UserName

Viewing profile as user:
SN

CAS RN 883-93-2

IUPAC Name: 2-phenyl-1,3-benzothiazole
Synonyms: 2-phenylbenzthiazole
Molecular Weight (g/mol): 211.28
Molecular Formula: C13H9NS
InChi Key: XBHOUXSGHYZCNH-UHFFFAOYSA-N
SMILES: S1C2=CC=CC=C2N=C1C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results