accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 883291-45-0

IUPAC Name: 1-[4-(2-methyl-1H-imidazol-1-yl)phenyl]methanamine
Synonyms: 1-[4-(2-methylimidazol-1-yl)phenyl]methanamine
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: JOMOQABDRBJMDQ-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=C(CN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results