accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrClF

CAS RN 883499-24-9

IUPAC Name: 1-bromo-2-chloro-3-fluorobenzene
Synonyms: 1-bromo-2-chloro-3-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: GTIWFNAUYPVPAT-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C(Br)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results