accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNFBr

CAS RN 883500-66-1

IUPAC Name: 4-bromo-7-fluoro-1H-indole
Synonyms: 4-bromo-7-fluoro-1H-indole
Molecular Weight (g/mol): 214.04
Molecular Formula: C8H5BrFN
InChi Key: PBQKMEMSAFAVKF-UHFFFAOYSA-N
SMILES: FC1=C2NC=CC2=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results