accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 883738-38-3

IUPAC Name: 1-methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Synonyms: 1-methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine
Molecular Weight (g/mol): 330.24
Molecular Formula: C18H27BN2O3
InChi Key: ZYDMNNQZEUSDGG-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C(=O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results