accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884495-39-0

IUPAC Name: 5-bromo-2-methoxypyridin-3-amine
Synonyms: 5-bromo-2-methoxypyridin-3-amine
Molecular Weight (g/mol): 203.04
Molecular Formula: C6H7BrN2O
InChi Key: HJOOFLFWIISCAI-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results