accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884507-16-8

IUPAC Name: 5-isocyanato-1-methyl-1H-indole
Synonyms: 5-isocyanato-1-methylindole
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: RDQRMCJCROUHOF-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC(=CC=C12)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results