accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3CH3CH3NH3C

CAS RN 884507-19-1

IUPAC Name: 1-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
Synonyms: 1-methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indole
Molecular Weight (g/mol): 257.14
Molecular Formula: C15H20BNO2
InChi Key: AYJLGLUJQKZRDL-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=C(C=C12)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results