accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884507-25-9

IUPAC Name: {2-[(1H-1,2,4-triazol-1-yl)methyl]phenyl}methanol
Synonyms: [2-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: OYFLAXVFVKLQPM-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results