accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884507-30-6

IUPAC Name: 2-(thiomorpholin-4-yl)pyridine-4-carbonitrile
Synonyms: 2-(thiomorpholin-4-yl)pyridine-4-carbonitrile
Molecular Weight (g/mol): 205.28
Molecular Formula: C10H11N3S
InChi Key: MZIANIKUVGCEPB-UHFFFAOYSA-N
SMILES: N#CC1=CC(=NC=C1)N1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results