accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884507-56-6

IUPAC Name: 1-[(3-iodophenyl)methyl]-1H-pyrazole
Synonyms: 1-[(3-iodophenyl)methyl]pyrazole
Molecular Weight (g/mol): 284.1
Molecular Formula: C10H9IN2
InChi Key: GWLQKUUXEMKUEW-UHFFFAOYSA-N
SMILES: IC1=CC=CC(CN2C=CC=N2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results