accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 884507-60-2

OONN

CAS RN 884507-60-2

IUPAC Name: 6-(oxan-4-yloxy)pyridine-3-carbonitrile
Synonyms: 6-(oxan-4-yloxy)pyridine-3-carbonitrile
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: CCEWHGWDICBASG-UHFFFAOYSA-N
SMILES: N#CC1=CN=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results