accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88493-55-4

IUPAC Name: 5,5'-bis({[2,2'-bithiophen]-5-yl})-2,2'-bithiophene
Synonyms: 5,5'-bis({[2,2'-bithiophen]-5-yl})-2,2'-bithiophene
Molecular Weight (g/mol): 494.74
Molecular Formula: C24H14S6
InChi Key: KUJYDIFFRDAYDH-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)