accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OON(S)OO

CAS RN 88495-54-9

IUPAC Name: (3S)-1-[(tert-butoxy)carbonyl]piperidine-3-carboxylate
Synonyms: (3S)-1-(tert-butoxycarbonyl)piperidine-3-carboxylate
Molecular Weight (g/mol): 228.27
Molecular Formula: C11H18NO4
InChi Key: NXILIHONWRXHFA-QMMMGPOBSA-M
SMILES: CC(C)(C)OC(=O)N1CCC[C@@H](C1)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)