accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 885266-56-8

IUPAC Name: tert-butyl N-[1-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-3-phenylpropan-2-yl]carbamate
Synonyms: tert-butyl N-[1-(1,3-dioxoisoindol-2-yl)-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 380.44
Molecular Formula: C22H24N2O4
InChi Key: ZBCKKSQEEBLGDU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CN1C(=O)C2=CC=CC=C2C1=O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results