accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNHNO

CAS RN 885266-77-3

IUPAC Name: tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate
Synonyms: tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate
Molecular Weight (g/mol): 260.29
Molecular Formula: C14H16N2O3
InChi Key: XKYBSNNENLRYPN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=C2C(NC=C2C=O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results