accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 885278-10-4

IUPAC Name: 2-(propan-2-yloxy)pyridine-3-carbaldehyde
Synonyms: 2-isopropoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: GNTQOKGIVMJHQG-UHFFFAOYSA-N
SMILES: CC(C)OC1=C(C=O)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results