accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 885278-12-6

IUPAC Name: 2-propoxypyridine-3-carbaldehyde
Synonyms: 2-propoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: CBTKKULPWZVYOL-UHFFFAOYSA-N
SMILES: CCCOC1=C(C=O)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results