accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 885280-64-8

IUPAC Name: N1,N1-dimethylcyclohexane-1,3-diamine
Synonyms: N1,N1-dimethylcyclohexane-1,3-diamine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: KKGNWXHXFXNADS-UHFFFAOYNA-N
SMILES: CN(C)C1CCCC(N)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results