accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88571-77-1

IUPAC Name: oxane-4-carbothioamide
Synonyms: oxane-4-carbothioamide
Molecular Weight (g/mol): 145.22
Molecular Formula: C6H11NOS
InChi Key: ZUONFEFOQCUTDW-UHFFFAOYSA-N
SMILES: NC(=S)C1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results