accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 885952-60-3

IUPAC Name: N'-hydroxy-2-(3-methylphenyl)ethanimidamide
Synonyms: N'-hydroxy-2-(3-methylphenyl)ethanimidamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: PUBSVUGAWXVFMA-UHFFFAOYSA-N
SMILES: CC1=CC(CC(N)=NO)=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results