accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 885960-32-7

H2NNHHOH2NNO(E/Z)(E/Z)

CAS RN 885960-32-7

IUPAC Name: 1-{6-[amino(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-ylidene}-1-nitrosomethanamine
Synonyms: 1-{6-[amino(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-ylidene}-1-nitrosomethanamine
Molecular Weight (g/mol): 194.19
Molecular Formula: C8H10N4O2
InChi Key: CQHMIKPVOMZRKW-UHFFFAOYSA-N
SMILES: NC(NO)=C1C=CC=CC1=C(N)N=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results