accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886-66-8

IUPAC Name: (4-phenylbuta-1,3-diyn-1-yl)benzene
Synonyms: diphenylbutadiyne diphenyldiacetylene
Molecular Weight (g/mol): 202.26
Molecular Formula: C16H10
InChi Key: HMQFJYLWNWIYKQ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C#CC#CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)