CAS RN 886456-80-0
CAS RN 886456-80-0
IUPAC Name:
2,7-bis(pyren-1-yl)-9,9'-spirobi[fluorene]
Synonyms:
2,7-bis(pyren-1-yl)-9,9'-spirobi[fluorene]
Molecular Weight (g/mol):
716.88
Molecular Formula:
C57H32
InChi Key:
MJXRNMQMLMXICM-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C1=CC=CC=C1C21C2=C(C=CC(=C2)C2=CC=C3C=CC4=CC=CC5=CC=C2C3=C45)C2=C1C=C(C=C2)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
1
–
1
of
1
results
1
2,7-Di(1-pyrenyl)-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™

CAS: 886456-80-0 Molecular Formula: C57H32 Molecular Weight (g/mol): 716.883 MDL Number: MFCD12022456 InChI Key: MJXRNMQMLMXICM-UHFFFAOYSA-N PubChem CID: 86242657 IUPAC Name: 2',7'-di(pyren-1-yl)-9,9'-spirobi[fluorene] SMILES: C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=C(C=CC(=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6)C1=C4C=C(C=C1)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1