accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886499-08-7

IUPAC Name: 2,4-difluoro-3-methoxyaniline
Synonyms: 2,4-difluoro-3-methoxyaniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: ISMLABQIDHXORP-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(N)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results