accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886531-62-0

IUPAC Name: (2-phenylpyrimidin-5-yl)methanol
Synonyms: (2-phenylpyrimidin-5-yl)methanol
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: DJEBODXMCJKLPH-UHFFFAOYSA-N
SMILES: OCC1=CN=C(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results