accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886766-60-5

IUPAC Name: tert-butyl (2R)-2-phenylpiperazine-1-carboxylate
Synonyms: tert-butyl (2R)-2-phenylpiperazine-1-carboxylate
Molecular Weight (g/mol): 262.35
Molecular Formula: C15H22N2O2
InChi Key: DVOURBIBCQYVCC-ZDUSSCGKSA-N
SMILES: CC(C)(C)OC(=O)N1CCNC[C@H]1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results