accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886791-73-7

IUPAC Name: 2-fluoro-N-[(4-fluorophenyl)methyl]aniline
Synonyms: 2-fluoro-N-[(4-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 219.24
Molecular Formula: C13H11F2N
InChi Key: KQBMUYYUEWLGQP-UHFFFAOYSA-N
SMILES: FC1=CC=C(CNC2=CC=CC=C2F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results