accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886851-29-2

IUPAC Name: ethyl 3-(1,3-thiazol-2-yl)benzoate
Synonyms: ethyl 3-(1,3-thiazol-2-yl)benzoate
Molecular Weight (g/mol): 233.29
Molecular Formula: C12H11NO2S
InChi Key: LHYDRQQHIJUWDE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results