accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 886851-42-9

IUPAC Name: 6-(furan-2-yl)pyridine-3-carbaldehyde
Synonyms: 6-(furan-2-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: RHHAYCIYWCFECO-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results