accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886851-44-1

IUPAC Name: {[6-(furan-2-yl)pyridin-3-yl]methyl}(methyl)amine
Synonyms: {[6-(furan-2-yl)pyridin-3-yl]methyl}(methyl)amine
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: UDZJNNURWGNFCN-UHFFFAOYSA-N
SMILES: CNCC1=CN=C(C=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results