accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886851-50-9

IUPAC Name: 2-(morpholin-4-yl)-5-(trifluoromethyl)benzaldehyde
Synonyms: 2-(morpholin-4-yl)-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 259.23
Molecular Formula: C12H12F3NO2
InChi Key: DIQGXZKPOSSQDH-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(C=O)=C(C=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results