accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 886851-52-1

IUPAC Name: methyl({[2-(morpholin-4-yl)-5-(trifluoromethyl)phenyl]methyl})amine
Synonyms: methyl({[2-(morpholin-4-yl)-5-(trifluoromethyl)phenyl]methyl})amine
Molecular Weight (g/mol): 274.29
Molecular Formula: C13H17F3N2O
InChi Key: UAZNBBVDDCKGES-UHFFFAOYSA-N
SMILES: CNCC1=C(C=CC(=C1)C(F)(F)F)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results