accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH2NO

CAS RN 886851-66-7

IUPAC Name: 3-amino-1-methyl-1H-pyrazole-4-carbaldehyde
Synonyms: 3-amino-1-methylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: UGWUYEXHMDPAMR-UHFFFAOYSA-N
SMILES: CN1C=C(C=O)C(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results