accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88686-29-7

IUPAC Name: (2-methoxy-5-methylphenyl)thiourea
Synonyms: 2-methoxy-5-methylphenylthiourea
Molecular Weight (g/mol): 196.27
Molecular Formula: C9H12N2OS
InChi Key: IPKUQBKZOAIUOC-UHFFFAOYSA-N
SMILES: COC1=C(NC(N)=S)C=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results