accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 887-54-7

IUPAC Name: 2,3,5,6-tetrachloro-4-(methoxycarbonyl)benzoic acid
Synonyms: 2,3,5,6-tetrachloro-4-(methoxycarbonyl)benzoic acid
Molecular Weight (g/mol): 317.93
Molecular Formula: C9H4Cl4O4
InChi Key: SXINVWXSZUQKSW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C(Cl)=C(C(O)=O)C(Cl)=C1Cl
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results