accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 887-76-3

HONNSOOO

CAS RN 887-76-3

IUPAC Name: 4-(diazyn-1-ium-1-yl)-3-hydroxynaphthalene-1-sulfonate
Synonyms: 4-(diazyn-1-ium-1-yl)-3-hydroxynaphthalene-1-sulfonate
Molecular Weight (g/mol): 250.23
Molecular Formula: C10H6N2O4S
InChi Key: QHIBNGIZPPHJAT-UHFFFAOYSA-N
SMILES: OC1=CC(=C2C=CC=CC2=C1[N+]#N)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results