accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOPOOFFFOFFF

CAS RN 88738-78-7

IUPAC Name: methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
Synonyms: methyl 2-[bis(2,2,2-trifluoroethoxy)phosphoryl]acetate
Molecular Weight (g/mol): 318.11
Molecular Formula: C7H9F6O5P
InChi Key: PVSJXEDBEXYLML-UHFFFAOYSA-N
SMILES: COC(=O)CP(=O)(OCC(F)(F)F)OCC(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)